4-Chlorothiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

A12021-09 A12021-18 A12021-14 A12021-22
CAAAAA12021-09EA 83.9 CAD
CAAAAA12021-09 CAAAAA12021-18 CAAAA12021-14 CAAAA12021-22
4-Chlorothiobenzamide 97%
4-Chlorothiobenzamide

MDL: MFCD00040956 Beilstein Registry No.: 606603


Purity: 97% CAS No.: 2521-24-6 Molecular Formula: C7H6ClNS Formula Weight: 171.65 Melting Point: 123-129° Beilstein Reference: 606603

Formula: C₇H₆ClNS
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 123…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00040956
CAS Number: 2521-24-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR