1-Octadecanol 97%
Supplier: Thermo Scientific Chemicals
A12020-30
A12020-0E
A12020-36
CAAAAA12020-30EA
51.67
CAD
CAAAAA12020-30
CAAAA12020-0E
CAAAA12020-36
1-Octadecanol 97%
1-Octadecanol
MDL: MFCD00002823
Beilstein Registry No.: 1362907
Purity: 97%
CAS No.: 112-92-5
Molecular Formula: CH3(CH2)17OH
Formula Weight: 270.50
Flash Point: 174°(345°F)
Physical Form: Flakes or pellets
Melting Point: 56-60°
Boiling Point: 210°/15mm
Beilstein Reference: 1362907
Merck Reference: 14,8805
MDL No.: MFCD00002823
Formula:
H₃C(CH₂)₁₆CH₂OH Boiling Pt: 210 °C/15 mm Melting Pt: 56…60 °C Density: 0.805 to 0.815 g/cm³ (60 °C) Flash Pt: 174 °C(345 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002823 CAS Number: 112-92-5 EINECS: 204-017-6 Merck Index: 14,8805 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...