3-Chloroaniline 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12019-22
A12019-36
CAAAAA12019-22EA
59.88
CAD
CAAAAA12019-22
CAAAA12019-36
3-Chloroaniline 99%
3-Chloroaniline
MDL: MFCD00007765
Beilstein Registry No.: 605969
Purity: 99%
CAS No.: 108-42-9
Molecular Formula: C6H6ClN
Formula Weight: 127.57
Flash Point: 123°(253°F)
Refractive Index: 1.5940
Odor: Characteristic
Melting Point: -10°
Boiling Point: 230°
Beilstein Reference: 605969
Merck Reference: 14,2118
UN No.: 2019
MDL No.: MFCD00007765
Formula:
C₆H₆ClN Boiling Pt: 230 °C Melting Pt: –10 °C Density: 1.210 Flash Pt: 123 °C (253 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007765 CAS Number: 108-42-9 EINECS: 203-581-0 UN: 2019 ADR: 6.1,II Merck Index: 14,2118 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...