N-(Chloromethyl)phthalimide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12018-14
A12018-22
CAAAA12018-14EA
162.74
CAD
CAAAA12018-14
CAAAA12018-22
N-(Chloromethyl)phthalimide 97%
N-(Chloromethyl)phthalimide
MDL: MFCD00005898
Beilstein Registry No.: 140942
Fieser: 1,135 18,100
Purity: 97%
CAS No.: 17564-64-6
Molecular Formula: C9H6ClNO2
Formula Weight: 195.61
Melting Point: 131-135°
Beilstein Reference: 140942
Formula:
C₉H₆ClNO₂ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005898 CAS Number: 17564-64-6 EINECS: 241-541-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...