4-Acetylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4-pyridyl ketone
A12015-14
A12015-22
CAAAA12015-14EA
66.52
CAD
CAAAA12015-14
CAAAA12015-22
CAAAA12015-36
4-Acetylpyridine 98%
4-Acetylpyridine
MDL: MFCD00006433
Beilstein Registry No.: 107629
Purity: 98%
CAS No.: 1122-54-9
Molecular Formula: C7H7NO
Formula Weight: 121.14
Flash Point: 104°(219°F)
Refractive Index: 1.5350
Melting Point: 14-16°
Boiling Point: 210-212°
Beilstein Reference: 107629
MDL No.: MFCD00006433
Formula:
C₇H₇NO Boiling Pt: 210…212 °C Melting Pt: 14…16 °C Density: 1.103 Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006433 CAS Number: 1122-54-9 EINECS: 214-350-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...