2-Chlorothiophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Chlorothiophenol, 2-chlorothiophenol, 1-Chloro-2-mercaptobenzene, 2-Chlorobenzenethiol, 2-Chlorobenzene-1-thiol

A12014-06 A12014-14 A12014-30
CAAAAA12014-06EA 58.78 CAD
CAAAAA12014-06 CAAAAA12014-14 CAAAA12014-30
2-Chlorothiophenol 98%
2-Chlorothiophenol

MDL: MFCD00004830 Beilstein Registry No.: 1209665


Purity: 98% CAS No.: 6320-03-2 Molecular Formula: C6H5ClS Formula Weight: 144.62 Flash Point: 88°(190°F) Refractive Index: 1.6040 Boiling Point: 108-110°/30mm Beilstein Reference: 1209665 UN No.: 1760 MDL No.: MFCD00004830

Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 108…110 °C (30 mmHg)
Density: 1.277
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004830
CAS Number: 6320-03-2
EINECS: 228-667-5
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR