1-Chloro-5-fluoro-2-iodobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12010-06
A12010-14
A12010-22
CAAAA12010-06EA
65.57
CAD
CAAAA12010-06
CAAAA12010-14
CAAAA12010-22
1-Chloro-5-fluoro-2-iodobenzene 98%
1-Chloro-5-fluoro-2-iodobenzene
MDL: MFCD00051774
Beilstein Registry No.: 2435080
Purity: 98%
CAS No.: 101335-11-9
Molecular Formula: C6H3ClFI
Formula Weight: 256.45
Refractive Index: 1.6010
Boiling Point: 96-98°/15mm
MDL No.: MFCD00051774
Formula:
C₆H₃ClFI Boiling Pt: 96…98 °C/15 mm Storage Temperature: Ambient |
MDL Number:
MFCD00051774 CAS Number: 101335-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...