4-Methyl-2-nitrophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Nitro-p-cresol, 1-Hydroxy-4-methyl-2-nitrobenzene

A12009-14 A12009-22 A12009-36
CAAAA12009-14EA 83.42 CAD
CAAAA12009-14 CAAAA12009-22 CAAAA12009-36
4-Methyl-2-nitrophenol 97%
4-Methyl-2-nitrophenol

MDL: MFCD00007120 Beilstein Registry No.: 1868022


Purity: 97% CAS No.: 119-33-5 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 108°(226°F) Odor: Phenol-like Boiling Point: 125°/22mm Beilstein Reference: 1868022 MDL No.: MFCD00007120

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 125 °C/22 mm
Melting Pt: 32…35 °C
Density: 1.240
Flash Pt: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007120
CAS Number: 119-33-5
EINECS: 204-315-6
UN: 2446
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR