1-Fluoro-4-iodobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Fluoroiodobenzene
A12008-09
A12008-18
A12008-30
CAAAA12008-09EA
52.14
CAD
CAAAA12008-09
CAAAA12008-18
CAAAA12008-30
1-Fluoro-4-iodobenzene 99%
1-Fluoro-4-iodobenzene
MDL: MFCD00001052
Beilstein Registry No.: 1853970
Purity: 99%
CAS No.: 352-34-1
Molecular Formula: C6H4FI
Formula Weight: 222.00
Flash Point: 68°(154°F)
Refractive Index: 1.5830
Melting Point: -20°
Boiling Point: 182-184°
Beilstein Reference: 1853970
MDL No.: MFCD00001052
Formula:
C₆H₄FI Boiling Pt: 182…184 °C Melting Pt: –20 °C Density: 1,925 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001052 CAS Number: 352-34-1 EINECS: 206-522-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...