Pyridine 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00011732
Beilstein Registry No.: 103233
Miscible with water, alcohol, ether, pet ether, oils
Fieser: 1,958 2,349 4,414 6,497 11,448 12,416
Purity: 99+%
CAS No.: 110-86-1
Molecular Formula: C5H5N
Formula Weight: 79.10
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 0.978
Refractive Index: 1.5100
Odor: Unpleasant
Melting Point: -42°
Boiling Point: 115°
Solubility: Miscible with water, alcohol, ether, pet ether, oils
Beilstein Reference: 103233
Merck Reference: 14,7970
UN No.: 1282
MDL No.: MFCD00011732
Formula:
C₅H₅N Boiling Pt: 115 °C Melting Pt: –42 °C Density: 0.978 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II Merck Index: 14,7970 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...