Tetrapropylammonium hydrogen sulfate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12002-06
A12002-14
CAAAA12002-03EA
67.34
CAD
CAAAA12002-03
CAAAA12002-06
CAAAA12002-14
Tetrapropylammonium hydrogen sulfate 98%
Tetrapropylammonium hydrogen sulphate
MDL: MFCD00038764
Purity: 98%
CAS No.: 56211-70-2
Molecular Formula: C12H29NO4S
Formula Weight: 283.43
Melting Point: 158-160°
MDL No.: MFCD00038764
Formula:
C₁₂H₂₉NO₄S Melting Pt: 158…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038764 CAS Number: 56211-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...