3-Chlorophenyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

A11999-06 A11999-14
CAAAA11999-06EA 63.52 CAD
CAAAA11999-06 CAAAA11999-14 CAAAA11999-22
3-Chlorophenyl isothiocyanate 97%
3-Chlorophenyl isothiocyanate

MDL: MFCD00004805 Beilstein Registry No.: 636856


Purity: 97% CAS No.: 2392-68-9 Molecular Formula: C7H4ClNS Formula Weight: 169.64 Flash Point: 110°(230°F) Refractive Index: 1.6585 Odor: Pungent Boiling Point: 249-250° Beilstein Reference: 636856 UN No.: 2922 MDL No.: MFCD00004805

Formula: C₇H₄ClNS
MW: 169.63
Boiling Pt: 249…250 °C
Melting Pt: 45
Density: 1.281
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004805
CAS Number: 2392-68-9
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR