3,4-Dimethylbenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

A11995-03 A11995-06 A11995-14
CAAAA11995-03EA 36.97 CAD
CAAAA11995-03 CAAAA11995-06 CAAAA11995-14
3,4-Dimethylbenzamide 98%
3,4-Dimethylbenzamide

MDL: MFCD00052960


Purity: 98% CAS No.: 5580-33-6 Molecular Formula: C9H11NO Formula Weight: 149.19 Melting Point: 101-103° MDL No.: MFCD00052960

Formula: C₉H₁₁NO
Melting Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00052960
CAS Number: 5580-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR