2-Deoxy-D(-)-ribose 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal
A11990-03
A11990-06
A11990-14
CAAAA11990-03EA
41.4
CAD
CAAAA11990-03
CAAAA11990-06
CAAAA11990-14
2-Deoxy-D(-)-ribose 99%
2-Deoxy-D(-)-ribose
MDL: MFCD00135904
Beilstein Registry No.: 1721978
Optical Rotation: -56°C (c=1 in water, 24h)
Purity: 99%
CAS No.: 533-67-5
Molecular Formula: C5H10O4
Formula Weight: 134.13
Melting Point: 76-85°
Beilstein Reference: 1721978
Merck Reference: 14,2908
Optical Rotation: -56° (c=1 in water, 24h)
MDL No.: MFCD00135904
Formula:
C₅H₁₀O₄ MW: 134.13 g/mol Boiling Pt: 292 °C (1013 hPa) Melting Pt: 76…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135904 CAS Number: 533-67-5 EINECS: 208-573-0 Merck Index: 14,2908 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...