2,5-Dimethyl-2,5-hexanediol 97%

Supplier: Thermo Scientific Chemicals
Danger

A11984-0E A11984-22 A11984-36
CAAAA11984-0EEA 497.7 CAD
CAAAA11984-0E CAAAA11984-22 CAAAA11984-36
2,5-Dimethyl-2,5-hexanediol 97%
2,5-Dimethyl-2,5-hexanediol

MDL: MFCD00004473 Beilstein Registry No.: 1361437


Purity: 97% CAS No.: 110-03-2 Molecular Formula: C8H18O2 Formula Weight: 146.23 Flash Point: 126°(258°F) Odor: Odorless Boiling Point: 214-215° Beilstein Reference: 1361437 MDL No.: MFCD00004473

Formula: C₈H₁₈O₂
Boiling Pt: 214…215 °C
Melting Pt: 87…91 °C
Density: 0.898
Flash Pt: 126 °C(258 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004473
CAS Number: 110-03-2
EINECS: 203-731-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR