2-Chloro-6-fluorobenzyl alcohol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Chloro-6-fluorophenyl)methanol
A11978-06
A11978-14
CAAAA11978-06EA
47.56
CAD
CAAAA11978-06
CAAAA11978-14
CAAAA11978-22
2-Chloro-6-fluorobenzyl alcohol 97%
2-Chloro-6-fluorobenzyl alcohol
MDL: MFCD00004608
Beilstein Registry No.: 1938353
Purity: 97%
CAS No.: 56456-50-9
Molecular Formula: C7H6ClFO
Formula Weight: 160.58
Flash Point:
Formula:
C₇H₆ClFO MW: 160.58 g/mol Boiling Pt: 232 °C (1013 hPa) Melting Pt: 42…44 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004608 CAS Number: 56456-50-9 EINECS: 260-192-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...