(±)-Phenylsuccinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A11971-14 A11971-22 A11971-36
CAAAA11971-14EA 46.14 CAD
CAAAA11971-14 CAAAA11971-22 CAAAA11971-36
(±)-Phenylsuccinic acid 98%
(±)-Phenylsuccinic acid

MDL: MFCD00004256 Beilstein Registry No.: 1875051


Purity: 98% CAS No.: 635-51-8 Molecular Formula: C10H10O4 Formula Weight: 194.19 Melting Point: 167-169° Beilstein Reference: 1875051

Formula: C₁₀H₁₀O₄
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 167…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00004256
CAS Number: 635-51-8
EINECS: 211-238-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR