2-Bromothiophene 98+%

Supplier: Thermo Scientific Chemicals
Danger

A11959-0B A11959-18 A11959-30
CAAAA11959-0BEA 774.2 CAD
CAAAA11959-0B CAAAA11959-18 CAAAA11959-30
2-Bromothiophene 98+%
2-Bromothiophene

MDL: MFCD00005417 Beilstein Registry No.: 104663


Purity: 98+% CAS No.: 1003-09-4 Molecular Formula: C4H3BrS Formula Weight: 163.04 Flash Point: 58°(136°F) Refractive Index: 1.5860 Melting Point: -10° Boiling Point: 149-152° Beilstein Reference: 104663 UN No.: 2929 MDL No.: MFCD00005417

Formula: C₄H₃BrS
Boiling Pt: 149…152 °C
Melting Pt: < –10 °C
Density: 1.701
Flash Pt: 58 °C(136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005417
CAS Number: 1003-09-4
EINECS: 213-699-4
UN: 2929
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR