Ethyl-5-bromovalerate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromovaleric acid ethyl ester, Ethyl 5-bromovalerate
A11952-14
A11952-22
A11952-36
CAAAA11952-14EA
121.34
CAD
CAAAA11952-14
CAAAA11952-22
CAAAA11952-36
Ethyl-5-bromovalerate 98%
Ethyl 5-bromovalerate
MDL: MFCD00000266
Beilstein Registry No.: 1752023
Purity: 98%
CAS No.: 14660-52-7
Molecular Formula: C7H13BrO2
Formula Weight: 209.09
Flash Point: 103°(217°F)
Refractive Index: 1.4580
Boiling Point: 227°
Beilstein Reference: 1752023
MDL No.: MFCD00000266
Formula:
BrH₂C(CH₂)₃COOCH₂CH₃ MW: 209.08 g/mol Boiling Pt: 227 °C Density: 1.306 Flash Pt: 103 °C(217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000266 CAS Number: 14660-52-7 EINECS: 238-705-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...