2,6-Xylenol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,6-Dimethylphenol, 2-Hydroxy-m-xylene, 2-Hydroxy-3-xylene

A11947-0E A11947-36
CAAAA11947-0EEA 325.48 CAD
CAAAA11947-0E CAAAA11947-36
2,6-Xylenol 99%
2,6-Xylenol

MDL: MFCD00002240 Beilstein Registry No.: 1446677


Purity: 99% CAS No.: 576-26-1 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 78°(172°F) Melting Point: 44-48° Boiling Point: 203° Beilstein Reference: 1446677 Merck Reference: 14,10082 UN No.: 2261 MDL No.: MFCD00002240

Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 203
Melting Pt: 44…48 °C
Density: 1.15
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002240
CAS Number: 576-26-1
EINECS: 209-400-1
UN: 2261
ADR: 6.1 II
Merck Index: 14,10082

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR