O-Phenylenediamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Phenylendiamine, 1,2-Phenylenediamine, 1,2-Diaminobenzene, OPD

A11946-14 A11946-0B A11946-0I A11946-30
CAAAAA11946-14EA 52.93 CAD
CAAAAA11946-14 CAAAA11946-0B CAAAA11946-0I CAAAA11946-30
O-Phenylenediamine 98%
o-Phenylenediamine

MDL: MFCD00007721 Beilstein Registry No.: 606074 Soluble in alcohol, chloroform, ether, slightly soluble in water Fieser: 1,834 8,393


Purity: 98% CAS No.: 95-54-5 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 136°(277°F) Physical Form: Crystals/flakes Density: 1.270 Sensitivity: Air and Light Sensitive Melting Point: 100-103° Boiling Point: 256-258° Solubility: Soluble in alcohol, chloroform, ether, slightly soluble in water Beilstein Reference: 606074 Merck Reference: 14,7284 UN No.: 1673 MDL No.: MFCD00007721

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 256…258 °C
Melting Pt: 100…103 °C
Density: 1.270
Flash Pt: 136 °C (277 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007721
CAS Number: 95-54-5
EINECS: 202-430-6
UN: 1673
ADR: 6.1 III
Merck Index: 14,7284

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR