4-Fluorophenol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11945-14
A11945-22
A11945-36
CAAAA11945-14EA
60.67
CAD
CAAAA11945-14
CAAAA11945-22
CAAAA11945-36
4-Fluorophenol 99%
4-Fluorophenol
MDL: MFCD00002316
Beilstein Registry No.: 1362752
Purity: 99%
CAS No.: 371-41-5
Molecular Formula: C6H5FO
Formula Weight: 112.10
Flash Point: 68°(154°F)
Odor: Phenol-like
Boiling Point: 185-188°
Beilstein Reference: 1362752
UN No.: 2923
MDL No.: MFCD00002316
Formula:
C₆H₅FO Boiling Pt: 185…188 °C Melting Pt: 44…50 °C Density: 1.22 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002316 CAS Number: 371-41-5 EINECS: 206-736-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...