3-Aminophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxyaniline, m-Aminophenol
A11943-0B
A11943-0I
A11943-30
CAAAA11943-0BEA
540.36
CAD
CAAAA11943-0B
CAAAA11943-0I
CAAAA11943-30
3-Aminophenol 98+%
3-Aminophenol
MDL: MFCD00007786
Beilstein Registry No.: 636059
Purity: 98+%
CAS No.: 591-27-5
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 178°(352°F)
Odor: Characteristic
pH: 6.8
Melting Point: 122-126°
Boiling Point: 164°/11mm
Beilstein Reference: 636059
Merck Reference: 14,460
UN No.: 2512
MDL No.: MFCD00007786
Formula:
C₆H₇NO MW: 109.13 g/mol Boiling Pt: 164 °C/11 mm Melting Pt: 122…126 °C Density: 0.99 Flash Pt: 178 °C (352 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007786 CAS Number: 591-27-5 EINECS: 209-711-2 UN: 2512 ADR: 6.1,III Merck Index: 14,460 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...