3-Bromobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11941-36
A11941-14
A11941-22
CAAAAA11941-36EA
745.76
CAD
CAAAAA11941-36
CAAAA11941-14
CAAAA11941-22
3-Bromobenzaldehyde 97%
3-Bromobenzaldehyde
MDL: MFCD00003345
Beilstein Registry No.: 1634534
Purity: 97%
CAS No.: 3132-99-8
Molecular Formula: C7H5BrO
Formula Weight: 185.03
Flash Point: 96°(204°F)
Refractive Index: 1.5940
Odor: Characteristic
Melting Point: 18-21°
Boiling Point: 234-236°
Beilstein Reference: 1634534
MDL No.: MFCD00003345
Formula:
C₇H₅BrO Boiling Pt: 234…236 °C Melting Pt: 18…21 °C Density: 1.580 Flash Pt: 96 °C(204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003345 CAS Number: 3132-99-8 EINECS: 221-526-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...