3-Bromo-2-thenoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11935-06
A11935-03
CAAAAA11935-06EA
161.16
CAD
CAAAAA11935-06
CAAAA11935-01
CAAAA11935-03
3-Bromo-2-thenoic acid 97%
3-Bromo-2-thenoic acid
MDL: MFCD00052291
Beilstein Registry No.: 121585
Purity: 97%
CAS No.: 7311-64-0
Molecular Formula: C5H3BrO2S
Formula Weight: 207.05
Melting Point: 195-196°
Beilstein Reference: 121585
Formula:
C₅H₃BrO₂S Melting Pt: 195…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052291 CAS Number: 7311-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...