4-Bromo-1,2-difluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11934-06
A11934-14
A11934-22
CAAAA11934-06EA
65.89
CAD
CAAAA11934-06
CAAAA11934-14
CAAAA11934-22
4-Bromo-1,2-difluorobenzene 98+%
4-Bromo-1,2-difluorobenzene
MDL: MFCD00000304
Beilstein Registry No.: 1934811
Purity: 98+%
CAS No.: 348-61-8
Molecular Formula: C6H3BrF2
Formula Weight: 192.99
Flash Point: 33°(91°F)
Refractive Index: 1.5050
Melting Point: -4°
Boiling Point: 150-151°
Beilstein Reference: 1934811
UN No.: 1993
MDL No.: MFCD00000304
Formula:
C₆H₃BrF₂ Boiling Pt: 150…151 °C Melting Pt: –4 °C Density: 1.707 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000304 CAS Number: 348-61-8 EINECS: 206-481-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...