1,3-Diaminopropane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11932-AE
A11932-0F
A11932-AP
CAAAAA11932-AEEA
50.24
CAD
CAAAAA11932-AE
CAAAA11932-0F
CAAAA11932-AP
1,3-Diaminopropane 98%
1,3-Diaminopropane
MDL: MFCD00008228
Beilstein Registry No.: 605277
Fieser: 13,160
Purity: 98%
CAS No.: 109-76-2
Molecular Formula: H2N(CH2)3NH2
Formula Weight: 74.13
Flash Point: 48°(118°F)
Refractive Index: 1.4573
Odor: Amine-like
pH:
Formula:
C₃H₁₀N₂ Boiling Pt: 134…136 °C Melting Pt: –12 °C Density: 0.888 Flash Pt: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008228 CAS Number: 109-76-2 EINECS: 203-702-7 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...