2-Thiophenecarbonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11930-36
A11930-14
A11930-22
CAAAAA11930-36EA
2782.38
CAD
CAAAAA11930-36
CAAAA11930-14
CAAAA11930-22
2-Thiophenecarbonitrile 98%
2-Thiophenecarbonitrile
MDL: MFCD00005416
Beilstein Registry No.: 107035
Purity: 98%
CAS No.: 1003-31-2
Molecular Formula: C5H3NS
Formula Weight: 109.15
Flash Point: 81°(178°F)
Refractive Index: 1.5645
Odor: Unpleasant
Boiling Point: 191-193°
Beilstein Reference: 107035
UN No.: 3276
MDL No.: MFCD00005416
Formula:
C₅H₃NS Boiling Pt: 191…193 °C Density: 1.171 Flash Pt: 81 °C (178 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005416 CAS Number: 1003-31-2 EINECS: 213-706-0 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...