2'-Methoxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11929-14
A11929-22
CAAAA11929-14EA
115.02
CAD
CAAAA11929-14
CAAAA11929-22
2'-Methoxyacetophenone 98%
2'-Methoxyacetophenone
MDL: MFCD00008725
Beilstein Registry No.: 1100602
Purity: 98%
CAS No.: 579-74-8
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 108°(226°F)
Refractive Index: 1.5393
Boiling Point: 119-120°/10mm
Beilstein Reference: 1100602
MDL No.: MFCD00008725
Formula:
C₉H₁₀O₂ Boiling Pt: 119…120 °C/10 mm Density: 1.090 Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008725 CAS Number: 579-74-8 EINECS: 209-446-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...