L(+)-Arabinose 99%
Supplier: Thermo Scientific Chemicals
A11921-18
A11921-30
CAAAA11921-18EA
137.3
CAD
CAAAA11921-18
CAAAA11921-30
L(+)-Arabinose 99%
L(+)-Arabinose
MDL: MFCD00067709
Beilstein Registry No.: 1723085
Optical Rotation: +104°C (c=10 in water, 24h)
Purity: 99%
CAS No.: 87-72-9
Molecular Formula: C5H10O5
Formula Weight: 150.13
Physical Form: Powder
Appearance: White
Odor: Odorless
Melting Point: 155-160°
Beilstein Reference: 1723085
Merck Reference: 14,761
Optical Rotation: +104° (c=10 in water, 24h)
MDL No.: MFCD00067709
Formula:
C₅H₁₀O₅ Boiling Pt: 331 °C (1013 hPa) Melting Pt: 155…160 °C Density: 1.585 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00067709 CAS Number: 5328-37-0 EINECS: 201-767-6 Merck Index: 14,761 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...