5-Bromosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-hydroxybenzaldehyde

A11917-14 A11917-22 A11917-36
CAAAA11917-14EA 98.43 CAD
CAAAA11917-14 CAAAA11917-22 CAAAA11917-36
5-Bromosalicylaldehyde 98%
5-Bromosalicylaldehyde

MDL: MFCD00003330 Beilstein Registry No.: 774710


Purity: 98% CAS No.: 1761-61-1 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Melting Point: 103-106° Beilstein Reference: 774710

Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00003330
CAS Number: 1761-61-1
EINECS: 217-167-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR