N,N-Dimethylaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

A11916-AE A11916-0D A11916-0F A11916-AP
CAAAAA11916-AEEA 42.66 CAD
CAAAAA11916-AE CAAAA11916-0D CAAAA11916-0F CAAAA11916-AP
N,N-Dimethylaniline 99%
N,N-Dimethylaniline

MDL: MFCD00008304 Beilstein Registry No.: 507140


Purity: 99% CAS No.: 121-69-7 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 62°(143°F) Physical Form: Liquid Density: 0.956 Refractive Index: 1.5580 Odor: Characteristic pH: 7.4 Melting Point: 2° Boiling Point: 193° Beilstein Reference: 507140 Merck Reference: 14,3234 UN No.: 2253 MDL No.: MFCD00008304

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 193 °C
Melting Pt: 2 °C
Density: 0.956
Flash Pt: 62 °C (143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008304
CAS Number: 121-69-7
EINECS: 204-493-5
UN: 2253
ADR: 6.1,II
Merck Index: 14,3234

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR