4-Chlorobenzhydrazide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chlorobenzoic acid hydrazide, 4-Chlorobenzohydrazide, 4-Chlorobenzene-1-carbohydrazide, 4-Chlorobenzoic hydrazide

A11911-18 A11911-06 A11911-22
CAAAAA11911-18EA 276.5 CAD
CAAAAA11911-18 CAAAA11911-06 CAAAA11911-22
4-Chlorobenzhydrazide 98%
4-Chlorobenzhydrazide

MDL: MFCD00007603 Beilstein Registry No.: 511154


Purity: 98% CAS No.: 536-40-3 Molecular Formula: C7H7ClN2O Formula Weight: 170.60 Melting Point: 162-166° Beilstein Reference: 511154

Formula: C₇H₇ClN₂O
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00007603
CAS Number: 536-40-3
EINECS: 208-632-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR