3-Buten-2-one tech. 90% stabilised, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Vinyl methyl ketone, MVK, Methyl vinyl ketone, Butenone
MDL: MFCD00008777
Beilstein Registry No.: 506021
Fieser: 1,697 2,283 5,464 6,407 7,247 10,272 12,329 13,193
Purity: 90%
CAS No.: 78-94-4
Molecular Formula: C4H6O
Formula Weight: 70.09
Storage Temperature: 2°C to 8°C
Flash Point: -6°(21°F)
Physical Form: Clear liquid
Density: 0.856
Refractive Index: 1.4120
Appearance: Colourless to pale yellow
Melting Point: -7°
Boiling Point: 32-34°/60mm
Beilstein Reference: 506021
Merck Reference: 14,6134
UN No.: 1251
MDL No.: MFCD00008777
Formula:
C₄H₆O MW: 70.09 Boiling Pt: 81.4 Melting Pt: -7 Density: 0.856 Flash Pt: –6 °C (21 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008777 CAS Number: 78-94-4 EINECS: 201-160-6 UN: 1251 ADR: 6.1 I Merck Index: 14,6134 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...