1-Iodododecane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Dodecyl iodide

Stabilization: Stabilised with copper

A11909-14 A11909-22 A11909-36
CAAAA11909-14EA 69.68 CAD
CAAAA11909-14 CAAAA11909-22 CAAAA11909-36
1-Iodododecane 98% stabilized
1-Iodododecane

MDL: MFCD00001088 Beilstein Registry No.: 1738943


Purity: 98% CAS No.: 4292-19-7 Molecular Formula: CH3(CH2)11I Formula Weight: 296.24 Flash Point: 110°(230°F) Refractive Index: 1.4844 Melting Point: -3° Boiling Point: 128-130°/11mm Beilstein Reference: 1738943 MDL No.: MFCD00001088

Formula: C₁₂H₂₅I
MW: 296.24 g/mol
Boiling Pt: 128…130 °C (11 mmHg)
Melting Pt: –3 °C
Density: 1.201
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001088
CAS Number: 4292-19-7
EINECS: 224-293-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR