Benzeneselenenyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylselenyl bromide, Phenyl hypobromoselenoite, Bromoselenobenzene, Phenylselenenyl bromide

A11906-03 A11906-06 A11906-14
CAAAAA11906-03EA 36.34 CAD
CAAAAA11906-03 CAAAA11906-06 CAAAA11906-14
Benzeneselenenyl bromide 98%
Benzeneselenenyl bromide

MDL: MFCD00000047 Beilstein Registry No.: 2039772 Fieser: 5,518 6,459 7,286 13,26 18,26,29 20,19


Purity: 98% CAS No.: 34837-55-3 Molecular Formula: C6H5BrSe Formula Weight: 235.98 Melting Point: 56-62° Boiling Point: 107-108°/15mm UN No.: 2928 MDL No.: MFCD00000047

Formula: (C₆H₅)SeBr
MW: 235.97 g/mol
Boiling Pt: 107…108 °C (15 mmHg)
Melting Pt: 56…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000047
CAS Number: 34837-55-3
EINECS: 252-238-1
UN: 2928
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR