α,2-Dichlorotoluene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Chlorobenzyl chloride, 1-Chloro-2-(chloromethyl)benzene
A11900-0E
A11900-36
CAAAA11900-0EEA
371.3
CAD
CAAAA11900-0E
CAAAA11900-36
α,2-Dichlorotoluene 98+%
α,2-Dichlorotoluene
MDL: MFCD00000893
Beilstein Registry No.: 471700
Purity: 98+%
CAS No.: 611-19-8
Molecular Formula: C7H6Cl2
Formula Weight: 161.03
Flash Point: 109°(228°F)
Refractive Index: 1.5590
Odor: Pungent
Melting Point: -13°
Boiling Point: 213-214°
Beilstein Reference: 471700
UN No.: 2235
MDL No.: MFCD00000893
Formula:
C₇H₆Cl₂ MW: 161.03 g/mol Boiling Pt: 213…214 °C Melting Pt: –13 °C Density: 1.272 Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000893 CAS Number: 611-19-8 EINECS: 210-258-8 UN: 2235 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...