1,4-Dichloro-2-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A11896-0E A11896-36
CAAAAA11896-30EA 159.85 CAD
CAAAAA11896-30 CAAAA11896-0E CAAAA11896-36
1,4-Dichloro-2-nitrobenzene 98%
1,4-Dichloro-2-nitrobenzene

MDL: MFCD00007074 Beilstein Registry No.: 778109


Purity: 98% CAS No.: 89-61-2 Molecular Formula: C6H3Cl2NO2 Formula Weight: 192.00 Flash Point: 125°(257°F) Melting Point: 53-57° Boiling Point: 266-269° Beilstein Reference: 778109 UN No.: 3077 MDL No.: MFCD00007074

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Boiling Pt: 266…269 °C
Melting Pt: 53…57 °C
Density: 1.6
Flash Pt: 125 °C(257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007074
CAS Number: 89-61-2
EINECS: 201-923-3
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR