Isopropyl cyanoacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isopropyl 2-cyanoacetate

A11887-14 A11887-22
CAAAA11887-14EA 61.62 CAD
CAAAA11887-14 CAAAA11887-22
Isopropyl cyanoacetate 97%
Isopropyl cyanoacetate

MDL: MFCD00019842 Beilstein Registry No.: 1756014


Purity: 97% CAS No.: 13361-30-3 Molecular Formula: NCCH2CO2CH(CH3)2 Formula Weight: 127.14 Flash Point: 92°(198°F) Refractive Index: 1.4150 Boiling Point: 207° Beilstein Reference: 1756014 MDL No.: MFCD00019842

Formula: C₆H₉NO₂
MW: 127.14 g/mol
Boiling Pt: 207 °C
Density: 1.013
Flash Pt: 92 °C (198 °F)
Storage Temperature: Ambient
MDL Number: MFCD00019842
CAS Number: 13361-30-3
EINECS: 236-421-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR