4-Biphenylylmethanol 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Biphenyl-4-methanol, [1,1'-Biphenyl]-4-ylmethanol, 4-Phenylbenzyl alcohol, 4-Biphenylmethanol

A11876-09 A11876-18 A11876-30
CAAAA11876-09EA 75.84 CAD
CAAAA11876-09 CAAAA11876-18 CAAAA11876-30
4-Biphenylylmethanol 98+%
4-Biphenylyl-methanol

MDL: MFCD00004660 Beilstein Registry No.: 1937761


Purity: 98+% CAS No.: 3597-91-9 Molecular Formula: C13H12O Formula Weight: 184.24 Melting Point: 98-104° Boiling Point: 184°/11mm MDL No.: MFCD00004660

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
Boiling Pt: 184 °C/11 mm
Melting Pt: 98…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00004660
CAS Number: 3597-91-9
EINECS: 222-745-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR