Sanger's reagent 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene

A11871-06 A11871-14 A11871-22
CAAAAA11871-06EA 66.36 CAD
CAAAAA11871-06 CAAAA11871-14 CAAAA11871-22
Sanger's reagent 99%
Sanger's reagent

MDL: MFCD00007056 Beilstein Registry No.: 398632 Fieser: 1,321 2,175 6,233


Purity: 99% CAS No.: 70-34-8 Molecular Formula: C6H3FN2O4 Formula Weight: 186.10 Flash Point: 110°(230°F) Refractive Index: 1.5690 Melting Point: 24-30° Boiling Point: 296° Beilstein Reference: 398632 Merck Reference: 14,4172 UN No.: 2811 MDL No.: MFCD00007056

Formula: C₆H₃FN₂O₄
MW: 186.1 g/mol
Boiling Pt: 296 °C
Melting Pt: 24…30 °C
Density: 1.482
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007056
CAS Number: 70-34-8
EINECS: 200-734-3
UN: 2811
ADR: 6.1,III
Merck Index: 14,4172

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR