Cumene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11864-AK
A11864-0F
A11864-AP
CAAAAA11864-AKEA
37.29
CAD
CAAAAA11864-AK
CAAAA11864-0F
CAAAA11864-AP
Cumene 99%
Cumene
MDL: MFCD00008881
Beilstein Registry No.: 1236613
Formula:
C₉H₁₂ Boiling Pt: 152…154 °C Melting Pt: –96 °C Density: 0.864 Flash Pt: 46 °C(114 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008881 CAS Number: 98-82-8 EINECS: 202-704-5 UN: 1918 ADR: "3",III Merck Index: 14,2617 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...