1-Naphthol 99%

Supplier: Thermo Scientific Chemicals
Danger

A11862-0E A11862-22 A11862-36
CAAAA11862-0EEA 488.22 CAD
CAAAA11862-0E CAAAA11862-22 CAAAA11862-36
1-Naphthol 99%
1-Naphthol

MDL: MFCD00003930 Beilstein Registry No.: 1817321 Soluble in alcohol, benzene, chloroform, ether


Purity: 99% CAS No.: 90-15-3 Molecular Formula: C10H8O Formula Weight: 144.17 Flash Point: 125°(257°F) Odor: schwach, phenolartig Boiling Point: 278-280° Solubility: Soluble in alcohol, benzene, chloroform, ether Beilstein Reference: 1817321 Merck Reference: 14,6383 UN No.: 2811 MDL No.: MFCD00003930

Formula: C₁₀H₈O
Boiling Pt: 278…280 °C
Melting Pt: 95…97 °C
Density: 1.28
Flash Pt: 125 °C (257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003930
CAS Number: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,6383

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR