4-tert-Butyl-α-chlorotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11856-06
A11856-14
CAAAA11856-06EA
101.12
CAD
CAAAA11856-06
CAAAA11856-14
4-tert-Butyl-α-chlorotoluene 99%
4-tert-Butyl-α-chlorotoluene
MDL: MFCD00000918
Beilstein Registry No.: 471673
Purity: 99%
CAS No.: 19692-45-6
Molecular Formula: C11H15Cl
Formula Weight: 182.70
Flash Point: 94° (202°F)
Refractive Index: 1.5220
Boiling Point: 101-104°/7mm
Beilstein Reference: 471673
UN No.: 1760
MDL No.: MFCD00000918
Formula:
C₁₁H₁₅Cl Boiling Pt: 101…104 °C/7 mm Density: 0.945 Flash Pt: 94°C (202 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000918 CAS Number: 19692-45-6 EINECS: 243-228-8 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...