Hexamethyldisiloxane, 0.65 cSt (25°C) ≥98%

Supplier: Thermo Scientific Chemicals
Danger

A11848-0F A11848-AE A11848-AP
CAAAA11848-0FEA 613.04 CAD
CAAAA11848-0F CAAAA11848-AE CAAAA11848-AP
Hexamethyldisiloxane, 0.65 cSt (25°C) ≥98%
Hexamethyldisiloxane

MDL: MFCD00008265 Beilstein Registry No.: 1736258 Fieser: 8,240 9,55 10,196 15,269 18,295


Purity: 98+% CAS No.: 107-46-0 Molecular Formula: C6H18OSi2 Formula Weight: 162.38 Flash Point: -2°(28°F) Refractive Index: 1.3770 Odor: Characteristic Melting Point: -67° Boiling Point: 100-101° Beilstein Reference: 1736258 UN No.: 1993 MDL No.: MFCD00008265

Formula: C₆H₁₈OSi₂
Boiling Pt: 100…101 °C
Melting Pt: –67 °C
Density: 0.764
Flash Pt: –2 °C (28 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008265
CAS Number: 107-46-0
EINECS: 203-492-7
UN: 1993
ADR: 3,III
REACH: 01-2119496108-31
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03d0d18c44dfa536ffcc853891b6f469333a38c5&vid=4959fefab2528efb6003e1f23c57966b65984db6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR