α,α,α-Trifluoro-3-iodotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α,α,α-Trifluoro-m-iodotoluene, 3-Iodobenzotrifluoride, 1-Iodo-3-(trifluoromethyl)benzene

A11843-09 A11843-18 A11843-30
CAAAA11843-09EA 47.4 CAD
CAAAA11843-09 CAAAA11843-18 CAAAA11843-30
α,α,α-Trifluoro-3-iodotoluene 98+%
α,α,α-Trifluoro-3-iodotoluene

MDL: MFCD00001049 Beilstein Registry No.: 2088596


Purity: 98+% CAS No.: 401-81-0 Molecular Formula: C7H4F3I Formula Weight: 272.01 Flash Point: 70°(158°F) Refractive Index: 1.5185 Melting Point: -8° Boiling Point: 81-82°/24mm Beilstein Reference: 2088596 MDL No.: MFCD00001049

Formula: C₇H₄F₃I
MW: 272.01 g/mol
Boiling Pt: 182
Melting Pt: -8
Density: 1.882
Flash Pt: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001049
CAS Number: 401-81-0
EINECS: 206-934-7
UN: 3265

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR