Tributyrin (glycerol tributyrate) 98%
Supplier: Thermo Scientific Chemicals
A11830-0B
A11830-18
A11830-30
CAAAA11830-0BEA
404.48
CAD
CAAAA11830-0B
CAAAA11830-18
CAAAA11830-30
Tributyrin (glycerol tributyrate) 98%
Tributyrin (Glycerol tributyrate)
A triglyceride that may inhibit cell growth and induce cell differentiation
Purity: 98%
CAS No.: 60-01-5
Molecular Formula: C15H26O6
Formula Weight: 302.37
Flash Point: 174°(345°F)
Refractive Index: 1.4358
Melting Point: -75°
Boiling Point: 307-308°
Beilstein Reference: 1714746
Merck Reference: 14,9620
MDL No.: MFCD00009392
Formula:
C₁₅H₂₆O₆ Boiling Pt: 307…308 °C Melting Pt: –75 °C Density: 1.032 Flash Pt: 174 °C(345 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009392 CAS Number: 60-01-5 EINECS: 200-451-5 Merck Index: 14,9620 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...