2-Methylresorcinol 98%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11821-0B
A11821-18
A11821-30
CAAAA11821-0BEA
771.04
CAD
CAAAA11821-0B
CAAAA11821-18
CAAAA11821-30
2-Methylresorcinol 98%, Technical Grade
2-Methylresorcinol
MDL: MFCD00002271
Beilstein Registry No.: 2042177
Purity: 98%
CAS No.: 608-25-3
Molecular Formula: C7H8O2
Formula Weight: 124.14
Flash Point: 135°(275°F)
Melting Point: 118-122°
Boiling Point: 264°
Beilstein Reference: 2042177
UN No.: 2811
MDL No.: MFCD00002271
Formula:
C₇H₈O₂ Boiling Pt: 264 °C Melting Pt: 118…122 °C Flash Pt: 135 °C (275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002271 CAS Number: 608-25-3 EINECS: 210-155-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...