2-Amino-4,6-dihydroxypyrimidine 98%

Supplier: Thermo Scientific Chemicals

A11813-14 A11813-22
CAAAA11813-14EA 84.21 CAD
CAAAA11813-14 CAAAA11813-22
2-Amino-4,6-dihydroxypyrimidine 98%
2-Amino-4,6-dihydroxypyrimidine

MDL: MFCD00006094 Beilstein Registry No.: 510297


Purity: 98% CAS No.: 56-09-7 Molecular Formula: C4H5N3O2 Formula Weight: 127.10 Melting Point: 300° Beilstein Reference: 510297

Formula: C₄H₅N₃O₂
Boiling Pt: 394 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006094
CAS Number: 56-09-7
EINECS: 200-256-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR