1-Chloro-2-fluoro-5-iodobenzene 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Chloro-1-fluoro-4-iodobenzene, 3-Chloro-4-fluoroiodobenzene

A11808-14 A11808-22
CAAAA11808-14EA 230.68 CAD
CAAAA11808-14 CAAAA11808-22
1-Chloro-2-fluoro-5-iodobenzene 98%
1-Chloro-2-fluoro-5-iodobenzene

MDL: MFCD00042210


Purity: 98% CAS No.: 156150-67-3 Molecular Formula: C6H3ClFI Formula Weight: 256.45 Flash Point: 110°(230°F) Refractive Index: 1.6050 Boiling Point: 94-95°/15mm MDL No.: MFCD00042210

Formula: C₆H₃ClFI
MW: 256.45
Boiling Pt: 94…95 °C/15 mm
Density: 2.020
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042210
CAS Number: 156150-67-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR